Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap2m1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201026.2 | NP_957320.1 | 6 - 7 | 886 | 60 | |
XM_005159256.4 | XP_005159313.1 | 7 - 8 | 937 | 60 | ||
GenBank mRNA | BC049515.1 | - | 6 - 7 | 888 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |