Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: u2af2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205689.1 | NP_991252.1 | 11 - 12 | 1063 | 73 | |
XM_009294031.2 | XP_009292306.1 | 12 - 13 | 1620 | 73 | ||
XM_009294032.3 | XP_009292307.1 | 12 - 13 | 1069 | 73 | ||
XM_009294033.3 | XP_009292308.1 | 11 - 12 | 1467 | 73 | ||
XM_009294034.3 | XP_009292309.1 | 11 - 12 | 1584 | 73 | ||
XM_009294035.3 | XP_009292310.1 | 10 - 11 | 1569 | 73 | ||
XM_009294036.3 | XP_009292311.1 | 10 - 11 | 1431 | 73 | ||
XM_009294037.3 | XP_009292312.1 | 9 - 10 | 977 | 73 | ||
GenBank mRNA | BC065869.1 | - | 11 - 12 | 1063 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |