Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207073.1 | NP_996956.1 | 8 - 9 | 932 | 69 | |
GenBank mRNA | BC066389.1 | - | 8 - 9 | 932 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |