Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207184.1 | NP_997067.1 | 1 - 2 | 419 | 61 | |
NM_207185.1 | NP_997068.1 | 1 - 2 | 407 | 61 | ||
GenBank mRNA | AF359505.1 | - | 1 - 2 | 419 | 61 | |
AY043241.1 | - | 1 - 2 | 407 | 61 | ||
BC092665.1 | - | 1 - 2 | 643 | 61 | ||
BC163896.1 | - | 1 - 2 | 297 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |