Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212595.1 | NP_997760.1 | 11 - 12 | 1377 | 67 | |
GenBank mRNA | AY394940.1 | - | 11 - 12 | 1377 | 67 | |
BC045315.1 | - | 11 - 12 | 1311 | 67 | ||
BC063984.1 | - | 11 - 12 | 1290 | 67 | ||
BC155193.1 | - | 11 - 12 | 1325 | 67 | ||
BC165845.1 | - | 11 - 12 | 1250 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |