Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213109.2 | NP_998274.2 | 1 - 2 | 289 | 95 | |
GenBank mRNA | BC049446.1 | - | 1 - 2 | 219 | 95 | |
EH555529.1 | - | 1 - 2 | 328 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |