Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213146.3 | NP_998311.2 | 6 - 7 | 1097 | 75 | |
GenBank mRNA | BC053246.1 | - | 6 - 7 | 1068 | 75 | |
EH562600.1 | - | 6 - 7 | 750 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |