Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214748.1 | NP_999913.1 | 6 - 7 | 432 | 84 | |
GenBank mRNA | BC065672.1 | - | 6 - 7 | 432 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |