Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnni4b.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002101.1 | NP_001002101.1 | 8 - 9 | 611 | 117 | |
XM_021470472.1 | XP_021326147.1 | 5 - 6 | 509 | 117 | ||
GenBank mRNA | AY929853.1 | - | 8 - 9 | 610 | 117 | |
BC071518.1 | - | 8 - 9 | 611 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |