Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002351.1 | NP_001002351.1 | 4 - 5 | 947 | 85 | |
GenBank mRNA | BC075908.1 | - | 4 - 5 | 947 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |