Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002592.1 | NP_001002592.1 | 3 - 4 | 348 | 78 | |
XM_005155627.4 | XP_005155684.1 | 4 - 5 | 596 | 78 | ||
GenBank mRNA | BC076006.1 | - | 3 - 4 | 348 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |