Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002592.1 | NP_001002592.1 | 4 - 5 | 487 | 98 | |
XM_005155627.4 | XP_005155684.1 | 5 - 6 | 735 | 98 | ||
GenBank mRNA | BC076006.1 | - | 4 - 5 | 487 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |