Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002593.2 | NP_001002593.1 | 2 - 3 | 339 | 100 | |
GenBank mRNA | BC076005.1 | - | 2 - 3 | 290 | 100 | |
EH448113.1 | - | 2 - 3 | 332 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |