Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab3db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002679.1 | NP_001002679.1 | 4 - 5 | 685 | 83 | |
XM_005168476.4 | XP_005168533.1 | 4 - 5 | 587 | 83 | ||
GenBank mRNA | BC075784.1 | - | 4 - 5 | 685 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |