Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suv39h1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003592.1 | NP_001003592.1 | 3 - 4 | 840 | 111 | |
GenBank mRNA | BC076417.1 | - | 3 - 4 | 840 | 111 | |
DQ840140.1 | - | 1 - 2 | 188 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |