Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suv39h1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003592.1 | NP_001003592.1 | 5 - 6 | 1113 | 91 | |
GenBank mRNA | BC076417.1 | - | 5 - 6 | 1113 | 91 | |
DQ840140.1 | - | 3 - 4 | 461 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Inventoried Probe spans exons |