Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdhr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005402.1 | NP_001005402.1 | 7 - 8 | 568 | 68 | |
XM_005156820.4 | XP_005156877.1 | 6 - 7 | 681 | 68 | ||
XM_021480567.1 | XP_021336242.1 | 7 - 8 | 702 | 68 | ||
GenBank mRNA | AY684930.1 | - | 7 - 8 | 568 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |