Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005601.2 | NP_001005601.1 | 3 - 4 | 327 | 121 | |
GenBank mRNA | BC082297.1 | - | 3 - 4 | 331 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |