Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlcd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005924.2 | NP_001005924.1 | 2 - 3 | 445 | 82 | |
GenBank mRNA | BC083537.1 | - | 2 - 3 | 445 | 82 | |
BC164600.1 | - | 1 - 2 | 177 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |