Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006091.1 | NP_001006091.1 | 4 - 5 | 441 | 69 | |
XM_021467085.1 | XP_021322760.1 | 6 - 7 | 1911 | 69 | ||
GenBank mRNA | BC083227.1 | - | 4 - 5 | 441 | 69 | |
BC153569.1 | - | 4 - 5 | 548 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |