Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vsnl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007366.2 | NP_001007367.1 | 2 - 3 | 392 | 76 | |
GenBank mRNA | BC085571.1 | - | 2 - 3 | 345 | 76 | |
BC164989.1 | - | 1 - 2 | 194 | 76 | ||
EH441888.1 | - | 2 - 3 | 306 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |