Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007415.1 | NP_001007416.1 | 13 - 14 | 1469 | 93 | |
XM_005171163.4 | XP_005171220.1 | 13 - 14 | 1330 | 93 | ||
XM_021480037.1 | XP_021335712.1 | 13 - 14 | 1448 | 93 | ||
GenBank mRNA | BC085370.1 | - | 13 - 14 | 1469 | 93 | |
BC165850.1 | - | 12 - 13 | 1248 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |