Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_155759.1 | 7 - 8 | 925 | 80 | ||
NR_155760.1 | 7 - 8 | 928 | 80 | |||
GenBank mRNA | BC091828.1 | - | 7 - 8 | 915 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |