Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014816.1 | NP_001014816.1 | 11 - 12 | 2627 | 97 | |
XM_005166379.4 | XP_005166436.1 | 12 - 13 | 2798 | 97 | ||
GenBank mRNA | AY954727.1 | - | 11 - 12 | 2627 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |