Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014816.1 | NP_001014816.1 | 13 - 14 | 2924 | 147 | |
XM_005166379.4 | XP_005166436.1 | 14 - 15 | 3095 | 147 | ||
GenBank mRNA | AY954727.1 | - | 13 - 14 | 2924 | 147 |
Gene Symbol: LOC103909932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009298287.3 | XP_009296562.2 | 2 - 3 | 334 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |