Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014816.1 | NP_001014816.1 | 6 - 7 | 1786 | 95 | |
XM_005166379.4 | XP_005166436.1 | 7 - 8 | 1957 | 95 | ||
GenBank mRNA | AY954727.1 | - | 6 - 7 | 1786 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |