Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017681.1 | NP_001017681.1 | 4 - 5 | 494 | 88 | |
GenBank mRNA | BC093160.1 | - | 4 - 5 | 494 | 88 | |
BC165257.1 | - | 4 - 5 | 371 | 88 |
Gene Symbol: LOC100536377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003201379.4 | XP_003201427.1 | 4 - 5 | 517 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |