Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fstl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017860.1 | NP_001017860.1 | 9 - 10 | 856 | 102 | |
GenBank mRNA | BC092990.1 | - | 9 - 10 | 856 | 102 | |
DQ317970.1 | - | 9 - 10 | 849 | 102 | ||
KM655580.1 | - | 8 - 9 | 800 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |