Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020510.1 | NP_001018346.1 | 5 - 6 | 2411 | 77 | |
XM_005160312.4 | XP_005160369.1 | 6 - 7 | 2659 | 77 | ||
GenBank mRNA | AY957463.1 | - | 5 - 6 | 2411 | 77 | |
BC171344.1 | - | 5 - 6 | 2464 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |