Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab6ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034179.1 | NP_001029351.1 | 6 - 7 | 619 | 78 | |
XM_005165932.4 | XP_005165989.1 | 6 - 7 | 648 | 78 | ||
GenBank mRNA | BC100011.1 | - | 6 - 7 | 619 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |