Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040687.1 | NP_001035777.1 | 9 - 10 | 1157 | 93 | |
XM_017351014.2 | XP_017206503.1 | 11 - 12 | 1749 | 93 | ||
XM_021481465.1 | XP_021337140.1 | 10 - 11 | 1633 | 93 | ||
GenBank mRNA | DQ466904.1 | - | 9 - 10 | 1157 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |