Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042754.1 | NP_001036219.1 | 3 - 4 | 553 | 110 | |
GenBank mRNA | BC095242.1 | - | 3 - 4 | 448 | 110 | |
BC117587.1 | - | 3 - 4 | 553 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |