Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pglyrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045222.2 | NP_001038687.2 | 4 - 5 | 1525 | 78 | |
GenBank mRNA | BC171349.1 | - | 4 - 5 | 1525 | 78 | |
DQ447204.1 | - | 4 - 5 | 1470 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |