Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045268.1 | NP_001038733.1 | 2 - 3 | 221 | 74 | |
XM_017355102.2 | XP_017210591.1 | 2 - 3 | 395 | 74 | ||
GenBank mRNA | BC116594.1 | - | 2 - 3 | 221 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |