Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glcci1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079950.1 | NP_001073419.1 | 3 - 4 | 980 | 114 | |
GenBank mRNA | AY835385.1 | - | 3 - 4 | 401 | 114 | |
BC129170.1 | - | 3 - 4 | 980 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |