Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amotl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080177.1 | NP_001073646.1 | 7 - 8 | 1861 | 82 | |
GenBank mRNA | BC163299.1 | - | 7 - 8 | 1735 | 82 | |
DQ887096.1 | - | 7 - 8 | 1861 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |