Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtac1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080178.1 | NP_001073647.1 | 11 - 12 | 1503 | 96 | |
XM_005160175.4 | XP_005160232.1 | 11 - 12 | 1585 | 96 | ||
XM_021475056.1 | XP_021330731.1 | 11 - 12 | 1586 | 96 | ||
XM_021475062.1 | XP_021330737.1 | 11 - 12 | 1554 | 96 | ||
XM_021475068.1 | XP_021330743.1 | 11 - 12 | 1584 | 96 | ||
GenBank mRNA | DQ351708.1 | - | 11 - 12 | 1503 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |