Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtac1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080178.1 | NP_001073647.1 | 3 - 4 | 450 | 91 | |
XM_005160175.4 | XP_005160232.1 | 3 - 4 | 532 | 91 | ||
XM_021475056.1 | XP_021330731.1 | 3 - 4 | 533 | 91 | ||
XM_021475062.1 | XP_021330737.1 | 3 - 4 | 501 | 91 | ||
XM_021475068.1 | XP_021330743.1 | 3 - 4 | 531 | 91 | ||
GenBank mRNA | BC146702.1 | - | 3 - 4 | 471 | 91 | |
DQ351708.1 | - | 3 - 4 | 450 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |