Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080607.2 | NP_001074076.2 | 7 - 8 | 1877 | 77 | |
XM_009305042.3 | XP_009303317.1 | 7 - 8 | 1944 | 77 | ||
XM_017357981.2 | XP_017213470.1 | 7 - 8 | 1947 | 77 | ||
XM_021479193.1 | XP_021334868.1 | 7 - 8 | 1952 | 77 | ||
XM_021479194.1 | XP_021334869.1 | 7 - 8 | 1953 | 77 | ||
GenBank mRNA | BC129173.1 | - | 7 - 8 | 1779 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |