Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082889.1 | NP_001076358.1 | 2 - 3 | 495 | 143 | |
GenBank mRNA | BC151857.1 | - | 2 - 3 | 521 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |