Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacnb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083555.1 | NP_001077024.1 | 4 - 5 | 509 | 80 | |
XM_005161994.4 | XP_005162051.1 | 4 - 5 | 680 | 80 | ||
XM_005161995.4 | XP_005162052.1 | 4 - 5 | 680 | 80 | ||
XM_017353586.2 | XP_017209075.1 | 4 - 5 | 680 | 80 | ||
GenBank mRNA | BC162364.1 | - | 4 - 5 | 488 | 80 | |
BC162649.1 | - | 4 - 5 | 488 | 80 | ||
DQ372946.1 | - | 4 - 5 | 509 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |