Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: terb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089382.1 | NP_001082851.1 | 12 - 13 | 1687 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |