Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: terb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089382.1 | NP_001082851.1 | 18 - 19 | 2397 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |