Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: terb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089382.1 | NP_001082851.1 | 2 - 3 | 243 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |