Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptmab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098730.1 | NP_001092200.1 | 1 - 2 | 143 | 79 | |
GenBank mRNA | BC059613.1 | - | 1 - 2 | 124 | 79 | |
BC142847.1 | - | 1 - 2 | 143 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |