Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: senp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021468556.1 | XP_021324231.1 | 20 - 21 | 2920 | 87 | |
XM_021468557.1 | XP_021324232.1 | 19 - 20 | 2856 | 87 | ||
XM_021468558.1 | XP_021324233.1 | 20 - 21 | 2907 | 87 | ||
XM_021468559.1 | XP_021324234.1 | 20 - 21 | 2917 | 87 | ||
XM_021468560.1 | XP_021324235.1 | 19 - 20 | 2826 | 87 | ||
XM_021468561.1 | XP_021324236.1 | 19 - 20 | 2813 | 87 | ||
GenBank mRNA | BC134226.1 | - | 4 - 5 | 384 | 87 | |
BC154420.1 | - | 7 - 8 | 929 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |