Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131049.1 | NP_571124.1 | 14 - 15 | 2164 | 96 | |
GenBank mRNA | AF073289.1 | - | 14 - 15 | 2164 | 96 | |
BC078280.1 | - | 13 - 14 | 1769 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |