Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131077.1 | NP_571152.1 | 2 - 3 | 140 | 80 | |
GenBank mRNA | BC115167.1 | - | 2 - 3 | 177 | 80 | |
X95301.1 | - | 140 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Probe spans exons |