Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rxrbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131238.1 | NP_571313.1 | 7 - 8 | 985 | 51 | |
XM_021466616.1 | XP_021322291.1 | 7 - 8 | 962 | 51 | ||
XM_021466617.1 | XP_021322292.1 | 7 - 8 | 964 | 51 | ||
GenBank mRNA | BC162301.1 | - | 7 - 8 | 979 | 51 | |
BC162302.1 | - | 7 - 8 | 979 | 51 | ||
U29941.1 | - | 7 - 8 | 985 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |