Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dacha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152955.2 | NP_694487.2 | 11 - 12 | 1923 | 105 | |
XM_005161406.4 | XP_005161463.1 | 11 - 12 | 1947 | 105 | ||
XM_009295592.3 | XP_009293867.1 | 11 - 12 | 1959 | 105 | ||
XM_009295593.3 | XP_009293868.1 | 11 - 12 | 1956 | 105 | ||
GenBank mRNA | AF427108.1 | - | 11 - 12 | 1924 | 105 | |
BC162471.1 | - | 11 - 12 | 1836 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |